Assay Details
| Gene Symbol: N6amt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500427.2 | XP_003500475.1 | 5 - 6 | 691 | 111 | |
| XM_007641167.1 | XP_007639357.1 | 5 - 6 | 931 | 111 | ||
| XM_007641168.1 | XP_007639358.1 | 5 - 6 | 815 | 111 | ||
| XM_007641169.1 | XP_007639359.1 | 5 - 6 | 662 | 111 | ||
| XM_007641170.1 | XP_007639360.1 | 5 - 6 | 846 | 111 | ||
| XM_007641171.1 | XP_007639361.1 | 4 - 5 | 556 | 111 | ||
| GenBank mRNA | JI879708.1 | - | 5 - 6 | 643 | 111 | |
More Information
Set Membership: |
|

N6amt2
Genomic Map