Assay Details
| Gene Symbol: Bsnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495753.1 | XP_003495801.1 | 3 - 4 | 522 | 69 | |
| XM_007643557.1 | XP_007641747.1 | 3 - 4 | 528 | 69 | ||
More Information
Set Membership: |
|

Bsnd
Genomic Map