Assay Details
| Gene Symbol: Tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502354.2 | XP_003502402.1 | 1 - 2 | 273 | 81 | |
| GenBank mRNA | JI880120.1 | - | 3 - 4 | 81 | ||
| JP054878.1 | - | 1 - 2 | 209 | 81 | ||
More Information
Set Membership: |
|

Tmed5
Genomic Map