Assay Details
| Gene Symbol: Snai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502472.2 | XP_003502520.1 | 1 - 2 | 206 | 66 | |
| XM_007643096.1 | XP_007641286.1 | 1 - 2 | 209 | 66 | ||
More Information
Set Membership: |
|

Snai1
Genomic Map