Assay Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506226.2 | XP_003506274.1 | 4 - 5 | 672 | 84 | |
| GenBank mRNA | JI894894.1 | - | 4 - 5 | 674 | 84 | |
| JP047933.1 | - | 4 - 5 | 674 | 84 | ||
More Information
Set Membership: |
|

Elavl1
Genomic Map