Assay Details
| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506507.2 | XP_003506555.1 | 2 - 3 | 351 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbox1| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506507.2 | XP_003506555.1 | 2 - 3 | 351 | 64 | |
Set Membership: |
|