Assay Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653952.1 | XP_007652142.1 | 6 - 7 | 585 | 63 | |
| XM_007653954.1 | XP_007652144.1 | 6 - 7 | 585 | 63 | ||
More Information
Set Membership: |
|

Chmp6
Genomic Map