Assay Details
| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645715.1 | XP_007643905.1 | 8 - 9 | 1548 | 63 | |
| GenBank mRNA | JI881162.1 | - | 8 - 9 | 1498 | 63 | |
| JP051156.1 | - | 8 - 9 | 1498 | 63 | ||
More Information
Set Membership: |
|

Leo1
Genomic Map