Assay Details
| Gene Symbol: Pde12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496102.2 | XP_003496150.1 | 2 - 3 | 1478 | 73 | |
| GenBank mRNA | JI883544.1 | - | 2 - 3 | 521 | 73 | |
| JP063586.1 | - | 2 - 3 | 520 | 73 | ||
More Information
Set Membership: |
|

Pde12
Genomic Map