Assay Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007633168.1 | XP_007631358.1 | 5 - 6 | 668 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ckb| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007633168.1 | XP_007631358.1 | 5 - 6 | 668 | 77 | |
Set Membership: |
|