Assay Details
| Gene Symbol: Nfkbia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650562.1 | XP_007648752.1 | 1 | 631 | 50 | |
| GenBank mRNA | JP053186.1 | - | 1 | 632 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfkbia| Gene Symbol: Nfkbia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650562.1 | XP_007648752.1 | 1 | 631 | 50 | |
| GenBank mRNA | JP053186.1 | - | 1 | 632 | 50 | |
Set Membership: |
|