Assay Details
| Gene Symbol: Rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652797.1 | XP_007650987.1 | 1 - 2 | 195 | 130 | |
| XM_007652798.1 | XP_007650988.1 | 1 - 2 | 157 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007652798.1 transcripts.
Set Membership: |
|

Rpl10
Genomic Map