Assay Details
| Gene Symbol: Arpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509682.2 | XP_003509730.1 | 7 - 8 | 1017 | 87 | |
| GenBank mRNA | JI878022.1 | - | 7 - 8 | 1020 | 87 | |
| JP037489.1 | - | 7 - 8 | 1020 | 87 | ||
More Information
Set Membership: |
|

Arpc1b
Genomic Map