Assay Details
| Gene Symbol: Ms4a6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653994.1 | XP_007652184.1 | 2 | 221 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a6a| Gene Symbol: Ms4a6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653994.1 | XP_007652184.1 | 2 | 221 | 77 | |
Set Membership: |
|