Assay Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515656.2 | XP_003515704.1 | 2 | 1121 | 93 | |
| XM_007607782.1 | XP_007605972.1 | 2 | 1047 | 93 | ||
| XM_007607783.1 | XP_007605973.1 | 2 | 1023 | 93 | ||
More Information
Set Membership: |
|

Ackr3
Genomic Map