Assay Details
| Gene Symbol: LOC103158893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608093.1 | XP_007606283.1 | 2 | 420 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103158893| Gene Symbol: LOC103158893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608093.1 | XP_007606283.1 | 2 | 420 | 64 | |
Set Membership: |
|