Assay Details
| Gene Symbol: IGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314299.1 | NP_001301228.1 | 3 - 4 | 794 | 54 | |
| GenBank mRNA | JP351124.2 | - | 1 - 2 | 509 | 54 | |
| JP351471.2 | - | 1 - 2 | 509 | 54 | ||
| JQ341160.1 | - | 3 - 4 | 794 | 54 | ||
More Information
Set Membership: |
|

IGFBP2
Genomic Map