Assay Details
| Gene Symbol: SDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054682.1 | XP_017910171.1 | 7 - 8 | 907 | 76 | |
| XM_018054683.1 | XP_017910172.1 | 6 - 7 | 838 | 76 | ||
| GenBank mRNA | JO632704.2 | - | 2 - 3 | 350 | 76 | |
| JQ692318.1 | - | 1 - 2 | 70 | 76 | ||
More Information
Set Membership: |
|

SDHB
Genomic Map