Assay Details
| Gene Symbol: EFNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056372.1 | XP_017911861.1 | 2 - 3 | 766 | 104 | |
| GenBank mRNA | JO428483.1 | - | 1 - 2 | 198 | 104 | |
More Information
Set Membership: |
|

EFNB2
Genomic Map