Assay Details
| Gene Symbol: RAB39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700559.3 | XP_005700616.2 | 1 - 2 | 510 | 95 | |
| GenBank mRNA | JO596684.2 | - | 1 - 2 | 156 | 95 | |
| JO637943.2 | - | 1 - 2 | 1022 | 95 | ||
More Information
Set Membership: |
|

RAB39B
Genomic Map