Assay Details
| Gene Symbol: CDK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065566.1 | XP_017921055.1 | 3 - 4 | 315 | 146 | |
| GenBank mRNA | JO595408.2 | - | 3 - 4 | 236 | 146 | |
More Information
Set Membership: |
|

CDK7
Genomic Map