Assay Details
| Gene Symbol: CKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066231.1 | XP_017921720.1 | 2 - 3 | 313 | 70 | |
| GenBank mRNA | KA352574.1 | - | 6 - 7 | 1678 | 70 | |
More Information
Set Membership: |
|

CKB
Genomic Map