Assay Details
| Gene Symbol: ENY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689152.3 | XP_005689209.1 | 4 - 5 | 384 | 101 | |
| XM_018058350.1 | XP_017913839.1 | 4 - 5 | 316 | 101 | ||
| GenBank mRNA | JO633255.2 | - | 1 - 2 | 170 | 101 | |
More Information
Set Membership: |
|

ENY2
Genomic Map