Assay Details
| Gene Symbol: ELAVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050993.1 | XP_017906482.1 | 4 - 5 | 582 | 81 | |
| GenBank mRNA | JO498968.2 | - | 2 - 3 | 789 | 81 | |
More Information
Set Membership: |
|

ELAVL1
Genomic Map