Assay Details
| Gene Symbol: IFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285682.1 | NP_001272611.1 | 3 - 4 | 366 | 73 | |
| GenBank mRNA | AY304501.1 | - | 3 - 4 | 366 | 73 | |
| AY603405.1 | - | 3 - 4 | 414 | 73 | ||
| EF375708.1 | - | 3 - 4 | 369 | 73 | ||
| U34232.1 | - | 3 - 4 | 418 | 73 | ||
More Information
Set Membership: |
|

IFNG
Genomic Map