Assay Details
| Gene Symbol: TSFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048002.1 | XP_017903491.1 | 5 - 6 | 668 | 65 | |
| GenBank mRNA | JO437927.2 | - | 5 - 6 | 683 | 65 | |
| JO629111.2 | - | 1 - 2 | 542 | 65 | ||
More Information
Set Membership: |
|

TSFM
Genomic Map