Assay Details
| Gene Symbol: GNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683799.3 | XP_005683856.1 | 3 - 4 | 1737 | 63 | |
| XM_018052092.1 | XP_017907581.1 | 3 - 4 | 695 | 63 | ||
More Information
Set Membership: |
|

GNA14
Genomic Map