Assay Details
| Gene Symbol: CCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285696.1 | NP_001272625.1 | 5 - 6 | 568 | 72 | |
| XM_018047538.1 | XP_017903027.1 | 4 - 5 | 473 | 72 | ||
| GenBank mRNA | AF506739.1 | - | 5 - 6 | 568 | 72 | |
More Information
Set Membership: |
|

CCNT1
Genomic Map