Assay Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695750.2 | XP_005695807.2 | 2 - 3 | 908 | 69 | |
| GenBank mRNA | JO633634.2 | - | 2 - 3 | 485 | 69 | |
More Information
Set Membership: |
|

ARL6IP5
Genomic Map