Assay Details
| Gene Symbol: PSMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678712.2 | XP_005678769.1 | 3 - 4 | 539 | 59 | |
| GenBank mRNA | JO637106.2 | - | 3 - 4 | 414 | 59 | |
More Information
Set Membership: |
|

PSMB2
Genomic Map