Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692819.3 | XP_005692876.2 | 4 - 5 | 709 | 63 | |
| XM_013971511.2 | XP_013826965.1 | 4 - 5 | 674 | 63 | ||
| XM_013971512.2 | XP_013826966.1 | 4 - 5 | 393 | 63 | ||
| GenBank mRNA | JO581400.1 | - | 4 - 5 | 795 | 63 | |
| JO581401.1 | - | 4 - 5 | 792 | 63 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map