Assay Details
| Gene Symbol: EIF3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682350.2 | XP_005682407.1 | 2 - 3 | 131 | 54 | |
| XM_013965149.1 | XP_013820603.1 | 2 - 3 | 131 | 54 | ||
More Information
Set Membership: |
|

EIF3G
Genomic Map