Assay Details
| Gene Symbol: UBE2V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058525.1 | XP_017914014.1 | 3 - 4 | 408 | 107 | |
| GenBank mRNA | JO413853.2 | - | 3 - 4 | 333 | 107 | |
| JO598064.2 | - | 1 - 2 | 1133 | 107 | ||
More Information
Set Membership: |
|

UBE2V2
Genomic Map