Assay Details
| Gene Symbol: PGK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700601.3 | XP_005700658.1 | 4 - 5 | 534 | 73 | |
| GenBank mRNA | JO376090.2 | - | 7 - 8 | 1281 | 73 | |
More Information
Set Membership: |
|

PGK1
Genomic Map