Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688961.3 | XP_005689018.1 | 2 - 3 | 733 | 72 | |
| GenBank mRNA | KA343475.1 | - | 2 - 3 | 745 | 72 | |
More Information
Set Membership: |
|

SNAI2
Genomic Map