Assay Details
| Gene Symbol: CHTOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677491.3 | XP_005677548.1 | 5 - 6 | 855 | 72 | |
| XM_018046140.1 | XP_017901629.1 | 5 - 6 | 852 | 72 | ||
| XM_018046141.1 | XP_017901630.1 | 3 - 4 | 465 | 72 | ||
| GenBank mRNA | KA334468.1 | - | 1 - 2 | 1239 | 72 | |
More Information
Set Membership: |
|

CHTOP
Genomic Map