Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692675.3 | XP_005692732.1 | 6 - 7 | 1096 | 61 | |
| XM_005692676.3 | XP_005692733.1 | 6 - 7 | 1095 | 61 | ||
| XM_005692677.3 | XP_005692734.1 | 5 - 6 | 902 | 61 | ||
| XM_013971514.2 | XP_013826968.1 | 5 - 6 | 992 | 61 | ||
More Information
Set Membership: |
|

TMEM143
Genomic Map