Assay Details
| Gene Symbol: POLR2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692272.3 | XP_005692329.1 | 3 - 4 | 620 | 68 | |
| XM_013971226.1 | XP_013826680.1 | 2 - 3 | 471 | 68 | ||
| GenBank mRNA | JO445792.2 | - | 3 - 4 | 569 | 68 | |
More Information
Set Membership: |
|

POLR2I
Genomic Map