Assay Details
| Gene Symbol: BLOC1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048122.1 | XP_017903611.1 | 3 - 4 | 496 | 70 | |
| GenBank mRNA | JO581386.1 | - | 3 - 4 | 411 | 70 | |
| KA337678.1 | - | 3 - 4 | 356 | 70 | ||
More Information
Set Membership: |
|

BLOC1S1
Genomic Map