Assay Details
| Gene Symbol: LEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682929.3 | XP_005682986.1 | 2 - 3 | 270 | 76 | |
| GenBank mRNA | JO438960.2 | - | 1 - 2 | 836 | 76 | |
More Information
Set Membership: |
|

LEAP2
Genomic Map