Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689120.3 | XP_005689177.1 | 2 - 3 | 359 | 57 | |
| XM_018058382.1 | XP_017913871.1 | 2 - 3 | 359 | 57 | ||
| GenBank mRNA | JO349574.2 | - | 3 - 4 | 765 | 57 | |
| JO418286.2 | - | 3 - 4 | 782 | 57 | ||
More Information
Set Membership: |
|

HEY1
Genomic Map