Assay Details
| Gene Symbol: MRPS23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693092.3 | XP_005693149.2 | 1 - 2 | 157 | 65 | |
| XM_018064100.1 | XP_017919589.1 | 1 - 2 | 162 | 65 | ||
| GenBank mRNA | JO393741.2 | - | 4 - 5 | 778 | 65 | |
| JO623489.2 | - | 1 - 2 | 174 | 65 | ||
More Information
Set Membership: |
|

MRPS23
Genomic Map