Assay Details
| Gene Symbol: ERI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678556.3 | XP_005678613.2 | 6 - 7 | 1007 | 57 | |
| XM_013962912.2 | XP_013818366.1 | 6 - 7 | 987 | 57 | ||
| XM_013962913.2 | XP_013818367.1 | 6 - 7 | 987 | 57 | ||
| XM_018043249.1 | XP_017898738.1 | 6 - 7 | 647 | 57 | ||
| XM_018043259.1 | XP_017898748.1 | 5 - 6 | 578 | 57 | ||
| XM_018043266.1 | XP_017898755.1 | 5 - 6 | 719 | 57 | ||
| XM_018043272.1 | XP_017898761.1 | 5 - 6 | 636 | 57 | ||
| XM_018043277.1 | XP_017898766.1 | 5 - 6 | 578 | 57 | ||
| XM_018043286.1 | XP_017898775.1 | 5 - 6 | 937 | 57 | ||
| XM_018043294.1 | XP_017898783.1 | 6 - 7 | 647 | 57 | ||
| XM_018043300.1 | XP_017898789.1 | 5 - 6 | 365 | 57 | ||
| GenBank mRNA | JO433541.2 | - | 6 - 7 | 879 | 57 | |
| JO622324.2 | - | 6 - 7 | 987 | 57 | ||
More Information
Set Membership: |
|

ERI3
Genomic Map