Assay Details
| Gene Symbol: UBE2Q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066095.1 | XP_017921584.1 | 9 - 10 | 1188 | 80 | |
| XM_018066096.1 | XP_017921585.1 | 9 - 10 | 1191 | 80 | ||
| GenBank mRNA | JO623778.2 | - | 9 - 10 | 1206 | 80 | |
More Information
Set Membership: |
|

UBE2Q2
Genomic Map