Assay Details
| Gene Symbol: MMGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976299.2 | XP_013831753.2 | 3 - 4 | 509 | 98 | |
| XM_018044237.1 | XP_017899726.1 | 3 - 4 | 265 | 98 | ||
| GenBank mRNA | JO615404.2 | - | 3 - 4 | 230 | 98 | |
| KA330571.1 | - | 1 | 365 | 98 | ||
More Information
Set Membership: |
|

MMGT1
Genomic Map