Assay Details
| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675023.2 | XP_005675080.1 | 1 - 2 | 122 | 76 | |
| GenBank mRNA | JO351622.2 | - | 1 - 2 | 45 | 76 | |
| JO546917.2 | - | 1 - 2 | 76 | 76 | ||
| JO593335.2 | - | 4 - 5 | 591 | 76 | ||
| KA346181.1 | - | 4 - 5 | 550 | 76 | ||
More Information
Set Membership: |
|

CCDC58
Genomic Map