Assay Details
| Gene Symbol: RAB5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693844.3 | XP_005693901.1 | 3 - 4 | 556 | 58 | |
| GenBank mRNA | JO340071.2 | - | 3 - 4 | 490 | 58 | |
More Information
Set Membership: |
|

RAB5C
Genomic Map