Assay Details
| Gene Symbol: EIF2S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700996.3 | XP_005701053.1 | 4 - 5 | 435 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF2S3| Gene Symbol: EIF2S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700996.3 | XP_005701053.1 | 4 - 5 | 435 | 60 | |
Set Membership: |
|