Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698752.3 | XP_005698809.3 | 1 - 2 | 457 | 79 | |
| XM_018042112.1 | XP_017897601.1 | 1 - 2 | 465 | 79 | ||
More Information
Set Membership: |
|

CDKN2AIP
Genomic Map