Assay Details
| Gene Symbol: N4BP2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056630.1 | XP_017912119.1 | 2 - 3 | 549 | 91 | |
| XM_018056631.1 | XP_017912120.1 | 2 - 3 | 556 | 91 | ||
| GenBank mRNA | JO408902.2 | - | 427 | 91 | ||
More Information
Set Membership: |
|

N4BP2L1
Genomic Map